Assay Details
Target Gene Details
Entrez Gene ID: | 83485 |
Gene Name: | neugrin, neurite outgrowth associated |
Gene Aliases: |
AW552001 |
Location: |
Chr.7:80261215-80265378 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ngrn | NM_031375.4 | NP_113552.3 | ||
NR_028053.1 | ||||
AB047544.1 | BAB21528.1 | |||
AK083330.1 | BAC38868.1 | |||
AK144756.1 | BAE26050.1 | |||
AK146059.1 | BAE26868.1 | |||
CJ154012.1 |
