Assay Details
Target Gene Details
Entrez Gene ID: | 14912 |
Gene Name: | NK6 homeobox 2 |
Gene Aliases: |
Gtx, Nkx6.2 |
Location: |
Chr.7:139579376-139582797 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx6-2 | NM_183248.3 | NP_899071.2 | ||
| NR_027857.1 | ||||
| AK008173.1 | BAC25203.1 | |||
| BY718566.1 | ||||
| L08074.1 |
More Information
Set Membership: |
|

Genomic Map