Assay Details
Target Gene Details
Entrez Gene ID: | 19943 |
Gene Name: | ribosomal protein L28 |
Gene Aliases: |
D7Wsu21e |
Location: |
Chr.7:4792888-4794562 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl28 | NM_009081.2 | NP_033107.1 | ||
| AK012368.1 | BAB28192.1 | |||
| AK018714.1 | BAB31362.1 | |||
| AK076133.1 | BAC36209.1 | |||
| BC024395.1 | AAH24395.1 | |||
| BC086797.1 | AAH86797.1 | |||
| BC086932.1 | AAH86932.1 | |||
| BC092012.1 | AAH92012.1 | |||
| X74856.1 | CAA52848.1 |
More Information
Set Membership: |
|

Genomic Map