Assay Details
Target Gene Details
Entrez Gene ID: | 67872 |
Gene Name: | non-SMC element 4 homolog A (S. cerevisiae) |
Gene Aliases: |
2410003A14Rik |
Location: |
Chr.7:130532526-130547381 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nsmce4a | NM_001162855.1 | NP_001156327.1 | ||
| AK010349.1 | ||||
| AK010795.1 | ||||
| AK042519.1 | 1 | 2187 | ||
| AK189457.1 | ||||
| BC089629.1 | AAH89629.1 |
More Information
Set Membership: |
|

Genomic Map