Assay Details
Target Gene Details
Entrez Gene ID: | 67186 |
Gene Name: | ribosomal protein, large P2 |
Gene Aliases: |
2700049I22Rik |
Location: |
Chr.7:141447464-141451350 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rplp2 | NM_026020.6 | NP_080296.3 | ||
| AK002419.1 | BAB22086.1 | |||
| AK008344.1 | BAB25616.1 | |||
| AK010618.1 | BAB27066.1 | |||
| AK012402.1 | BAB28217.1 | |||
| AK012526.1 | BAB28297.1 | |||
| AK028140.1 | BAC25768.1 | |||
| AK028149.1 | BAC25777.1 | |||
| AK088737.1 | BAC40539.1 | |||
| AK134206.1 | BAE22051.1 | |||
| AK166774.1 | BAE39010.1 | |||
| BC012413.1 | AAH12413.1 |
More Information
Set Membership: |
|

Genomic Map