Assay Details
Target Gene Details
Entrez Gene ID: | 210045 |
Gene Name: | NLR family, pyrin domain containing 4B |
Gene Aliases: |
AU022726, E330024M09, Nalp-gamma, Nalp4b |
Location: |
Chr.7:10687793-10730168 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nlrp4b | NM_172481.2 | 7 | 2395 | NP_766069.1 |
AK054426.1 | 7 | 2396 | BAC35775.1 | |
AK162217.1 | 4 | 1127 | BAE36795.1 | |
AY355342.1 | 7 | 2396 | AAR14739.1 | |
AY596198.1 | 5 | 2231 | AAU06319.1 | |
BC066795.1 | 7 | 2405 | AAH66795.1 |
