Assay Details
Target Gene Details
Entrez Gene ID: | 27050 |
Gene Name: | ribosomal protein S3 |
Gene Aliases: |
D7Ertd795e, Rs_3 |
Location: |
Chr.7:99477896-99483709 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rps3 | NM_012052.2 | NP_036182.1 | ||
AK003181.1 | BAB22624.1 | |||
AK010579.1 | BAB27042.1 | |||
AK010678.1 | ||||
AK011655.1 | BAB27761.1 | |||
AK012234.1 | BAB28111.1 | |||
AK012314.1 | BAB28159.1 | |||
AK012538.1 | ||||
AK051243.1 | BAC34570.1 | |||
AK088038.1 | ||||
AK143603.1 | ||||
AK146165.1 | BAE26946.1 | |||
AK146796.1 | BAE27438.1 | |||
AK148030.1 | BAE28299.1 | |||
AK150479.1 | BAE29595.1 | |||
AK159508.1 | BAE35140.1 | |||
AK160526.1 | BAE35844.1 | |||
AK164077.1 | BAE37615.1 | |||
AK165174.1 | BAE38059.1 | |||
AK166750.1 | BAE38991.1 | |||
AK167210.1 | BAE39337.1 | |||
AK167266.1 | BAE39379.1 | |||
AK168845.1 | BAE40668.1 | |||
AK206775.1 | ||||
BC010721.1 | AAH10721.1 | |||
CJ074413.1 | ||||
U89414.1 | AAC36521.1 | |||
X76772.1 | CAA54167.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
