Assay Details
Target Gene Details
Entrez Gene ID: | 17136 |
Gene Name: | myelin-associated glycoprotein |
Gene Aliases: |
Gma, siglec-4a |
Location: |
Chr.7:30899175-30914922 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mag | NM_010758.2 | NP_034888.1 | ||
BC101944.1 | AAI01945.1 | |||
BC103521.1 | AAI03522.1 | |||
BC103522.1 | AAI03523.1 | |||
BC103523.1 | AAI03524.1 | |||
M31811.1 | AAA39487.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
