Assay Details
Target Gene Details
Entrez Gene ID: | 77613 |
Gene Name: | protease, serine 36 |
Gene Aliases: |
C330007D15Rik |
Location: |
Chr.7:127932638-127946725 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss36 | NM_001081374.1 | NP_001074843.2 | ||
| AK021185.1 |
More Information
Set Membership: |
|

Genomic Map