Assay Details
Target Gene Details
Entrez Gene ID: | 68458 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 14A |
Gene Aliases: |
1110001M11Rik, Cpi17 |
Location: |
Chr.7:29289320-29293390 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r14a | NM_026731.3 | 4 | 469 | NP_081007.2 |
| AF352573.1 | 4 | 477 | AAK35214.1 | |
| AK003249.1 | 4 | 467 | ||
| AY050672.1 | 4 | 468 | AAL25830.1 | |
| BC010832.1 | 4 | 469 | AAH10832.1 |
More Information
Set Membership: |
|

Genomic Map