Assay Details
Target Gene Details
Entrez Gene ID: | 20174 |
Gene Name: | RuvB-like protein 2 |
Gene Aliases: |
mp47, p47 |
Location: |
Chr.7:45421896-45435170 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ruvbl2 | NM_011304.3 | 9 | 930 | NP_035434.1 |
AB013912.1 | 9 | 761 | BAA76297.1 | |
AK135404.1 | 9 | 744 | BAE22521.1 | |
AK144156.1 | 9 | 752 | BAE25736.1 | |
AK159112.1 | 9 | 932 | BAE34828.1 | |
AK189162.1 | ||||
AK191656.1 | ||||
AK203193.1 | ||||
AK205364.1 | ||||
AK210146.1 | ||||
AK212334.1 | ||||
AK212947.1 | ||||
BC115809.1 | 9 | 748 | ||
BC116693.1 | 9 | 747 |
