Assay Details
Target Gene Details
Entrez Gene ID: | 20266 |
Gene Name: | sodium channel, voltage-gated, type I, beta |
Gene Aliases: |
- |
Location: |
Chr.7:31116524-31126945 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scn1b | NM_011322.2 | 2 | 366 | NP_035452.1 |
| BC009652.1 | 2 | 366 | AAH09652.1 | |
| BC039140.1 | 3 | 369 | AAH39140.1 | |
| U46681.1 | 2 | 144 | AAC53006.1 | |
| U85786.1 | 2 | 186 | AAB49368.1 |
More Information
Set Membership: |
|

Genomic Map