Assay Details
Target Gene Details
Entrez Gene ID: | 20266 |
Gene Name: | sodium channel, voltage-gated, type I, beta |
Gene Aliases: |
- |
Location: |
Chr.7:31116524-31126945 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scn1b | NM_011322.2 | NP_035452.1 | ||
| AK028238.1 | 1 | 103 | BAC25833.1 | |
| BC009652.1 | AAH09652.1 | |||
| BC039140.1 | AAH39140.1 | |||
| U46681.1 | AAC53006.1 | |||
| U85786.1 | AAB49368.1 |
More Information
Set Membership: |
|

Genomic Map