Assay Details
Target Gene Details
Entrez Gene ID: | 170460 |
Gene Name: | StAR-related lipid transfer (START) domain containing 5 |
Gene Aliases: |
2310058G22Rik, AI607880, D7Ertd152e |
Location: |
Chr.7:83632017-83642328 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stard5 | NM_023377.4 | NP_075866.2 | ||
| AF480302.1 | AAL89652.1 | |||
| AK009980.1 | BAB26624.2 | |||
| AK077328.1 | BAC36753.1 | |||
| AK080060.1 | BAC37819.1 | |||
| AK085852.1 | BAC39551.1 | |||
| AK142503.1 | BAE25090.1 | |||
| AK158178.1 | BAE34395.1 | |||
| AY007808.1 | AAG41056.1 | |||
| BC058226.1 | AAH58226.1 |

Genomic Map