Assay Details
Target Gene Details
Entrez Gene ID: | 22282 |
Gene Name: | upstream transcription factor 2 |
Gene Aliases: |
Usf-2, bHLHb12 |
Location: |
Chr.7:30945248-30957250 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Usf2 | NM_011680.2 | NP_035810.1 | ||
AB221562.1 | ||||
AK146871.1 | BAE27494.1 | |||
AK158906.1 | ||||
AK211311.1 | ||||
BC082995.1 | AAH82995.1 | |||
U01662.1 | AAA20492.1 | |||
U01663.1 | AAA20493.1 |
