Assay Details
Target Gene Details
Entrez Gene ID: | 170460 |
Gene Name: | StAR-related lipid transfer (START) domain containing 5 |
Gene Aliases: |
2310058G22Rik, AI607880, D7Ertd152e |
Location: |
Chr.7:83632017-83642328 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stard5 | NM_023377.4 | 4 | 369 | NP_075866.2 |
AF480302.1 | 4 | 345 | AAL89652.1 | |
AK009980.1 | 4 | 375 | BAB26624.2 | |
AK077328.1 | 4 | 387 | BAC36753.1 | |
AK080060.1 | 4 | 387 | BAC37819.1 | |
AK085852.1 | 4 | 382 | BAC39551.1 | |
AK142503.1 | 4 | 387 | BAE25090.1 | |
AK158178.1 | 4 | 371 | BAE34395.1 | |
AY007808.1 | 4 | 364 | AAG41056.1 | |
BC058226.1 | 4 | 369 | AAH58226.1 |
