Assay Details
Target Gene Details
Entrez Gene ID: | 232976 |
Gene Name: | zinc finger protein 574 |
Gene Aliases: |
A630056B21Rik, Znf574 |
Location: |
Chr.7:25075684-25083492 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp574 | NM_001168506.1 | NP_001161978.1 | ||
| NM_175477.4 | NP_780686.1 | |||
| AK042074.1 | BAC31155.1 | |||
| AK053744.1 | BAC35502.1 | |||
| AK143997.1 | BAE25656.1 | |||
| AK170243.1 | BAE41657.1 | |||
| BC059044.1 | AAH59044.1 | |||
| BY087218.1 |
More Information
Set Membership: |
|

Genomic Map