Assay Details
Target Gene Details
Entrez Gene ID: | 66411 |
Gene Name: | tubulin folding cofactor B |
Gene Aliases: |
2410007D12Rik, AU041393, CG22, CKAPI, Ckap1 |
Location: |
Chr.7:30224130-30232029 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbcb | NM_025548.3 | NP_079824.2 | ||
| AK002316.1 | BAB22009.1 | |||
| AK003655.2 | BAB22918.2 | |||
| AK010438.1 | BAB26939.1 | |||
| AK183833.1 | ||||
| AK214432.1 | ||||
| BC010684.1 | AAH10684.1 |
More Information
Set Membership: |
|

Genomic Map