Assay Details
Target Gene Details
Entrez Gene ID: | 21990 |
Gene Name: | tryptophan hydroxylase 1 |
Gene Aliases: |
Tph |
Location: |
Chr.7:46644641-46672537 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tph1 | NM_001136084.2 | 6 | 668 | NP_001129556.1 |
NM_009414.3 | 6 | 979 | NP_033440.1 | |
AF273257.1 | 6 | 669 | AAF86975.1 | |
AK146134.1 | 6 | 668 | BAE26925.1 | |
AK146171.1 | 6 | 668 | BAE26950.1 | |
BC072582.1 | 6 | 979 | AAH72582.1 | |
J04758.1 | 6 | 639 | AAA63401.1 |
