Assay Details
Target Gene Details
Entrez Gene ID: | 17173 |
Gene Name: | achaete-scute complex homolog 2 (Drosophila) |
Gene Aliases: |
2410083I15Rik, Mash2, bHLHa45 |
Location: |
Chr.7:142966819-143009052 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ascl2 |
Target Gene Details
Entrez Gene ID: | 27027 |
Gene Name: | tetraspanin 32 |
Gene Aliases: |
AW208513, Art-1, BB235973, D7Wsu37e, Phemx, Tssc6 |
Location: |
Chr.7:143004902-143019486 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tspan32 | NM_001128080.2 | NP_001121552.1 | ||
NM_001128081.1 | NP_001121553.1 | |||
NM_001128082.1 | NP_001121554.1 | |||
NM_020286.3 | NP_064682.1 | |||
AF175771.1 | AAG27268.1 | |||
AF291425.1 | AAK83110.1 | |||
AJ279791.1 | CAB94719.1 | |||
AJ279792.1 | CAB94720.1 | |||
AJ279793.1 | CAB94721.1 | |||
AJ279794.1 | CAB94722.1 | |||
AJ279795.1 | CAB94723.1 | |||
AK013248.1 | ||||
AK017989.1 | 1 | 1351 | ||
AK040164.1 | 1 | 119 | ||
AK041748.1 | 3 | 363 | BAC31052.1 | |
AK154694.1 | BAE32769.1 | |||
AK171199.1 | ||||
AY039000.1 | AAK84430.1 | |||
BC055858.1 | AAH55858.1 |
