Assay Details
Target Gene Details
Entrez Gene ID: | 22228 |
Gene Name: | uncoupling protein 2 (mitochondrial, proton carrier) |
Gene Aliases: |
Slc25a8 |
Location: |
Chr.7:100493358-100499630 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ucp2 | NM_011671.5 | 5 | 840 | NP_035801.3 |
AF111998.1 | 3 | 543 | AAD17198.1 | |
AF111999.1 | 3 | 537 | AAD17199.1 | |
AK134116.1 | 5 | 841 | BAE22021.1 | |
AK142687.1 | 5 | 841 | BAE25160.1 | |
AK165245.1 | 5 | 841 | ||
BC012697.1 | 5 | 839 | AAH12697.1 | |
BC012967.1 | 4 | 741 | AAH12967.1 | |
U69135.1 | 5 | 768 | AAB17666.1 | |
U94593.1 | 5 | 761 | AAB53092.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
