Assay Details
Target Gene Details
Entrez Gene ID: | 71984 |
Gene Name: | seryl-aminoacyl-tRNA synthetase 2 |
Gene Aliases: |
2410015F05Rik, D7Ertd353e, SerRS, SerRSmt |
Location: |
Chr.7:28741934-28753879 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sars2 | NM_023637.3 | NP_076126.2 | ||
| AB029949.1 | BAA99558.1 | |||
| AK010491.1 | BAB26981.1 | |||
| AK087606.1 | BAC39943.1 | |||
| AK140760.1 | BAE24469.1 | |||
| AK178578.1 | ||||
| BC079664.1 | AAH79664.1 |
More Information
Set Membership: |
|

Genomic Map