Assay Details
Target Gene Details
Entrez Gene ID: | 68278 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 39 |
Gene Aliases: |
2610307C23Rik, BAT1, DDXL, Ddx39a, URH49 |
Location: |
Chr.8:83715177-83723351 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx39 | NM_197982.3 | NP_932099.2 | ||
AK011285.1 | ||||
AK012002.1 | ||||
AK016960.1 | ||||
AK145927.1 | BAE26758.1 | |||
AK146294.1 | BAE27051.1 | |||
AK149475.1 | ||||
AK150997.1 | BAE30021.1 | |||
AK151395.1 | BAE30363.1 | |||
AK152394.1 | BAE31182.1 | |||
AK164289.1 | BAE37719.1 | |||
AK167890.1 | BAE39902.1 | |||
AK168079.1 | BAE40052.1 | |||
AK172448.1 | BAE43010.1 | |||
AK187116.1 | ||||
BC020134.1 | AAH20134.1 | |||
CF172497.1 |
