Assay Details
Target Gene Details
Entrez Gene ID: | 66965 |
Gene Name: | cytosolic thiouridylase subunit 2 homolog (S. pombe) |
Gene Aliases: |
2310061F22Rik, C16orf84, Ncs2 |
Location: |
Chr.8:122476137-122483092 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctu2 | NM_153775.2 | NP_722470.2 | ||
| AK012440.1 | ||||
| AK090349.1 | BAC41181.1 | |||
| AK154995.1 | BAE32982.1 | |||
| AK172353.1 | ||||
| AK188865.1 | ||||
| AK206078.1 | ||||
| AK215943.1 | ||||
| BC036332.1 | AAH36332.1 |
More Information
Set Membership: |
|

Genomic Map