Assay Details
Target Gene Details
Entrez Gene ID: | 66965 |
Gene Name: | cytosolic thiouridylase subunit 2 homolog (S. pombe) |
Gene Aliases: |
2310061F22Rik, C16orf84, Ncs2 |
Location: |
Chr.8:122476137-122483092 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctu2 | NM_153775.2 | NP_722470.2 | ||
| AK012440.1 | ||||
| AK154995.1 | BAE32982.1 | |||
| AK172353.1 | 2 | 737 | ||
| BC036332.1 | AAH36332.1 | |||
| CJ067802.1 |
More Information
Set Membership: |
|

Genomic Map