Assay Details
Target Gene Details
Entrez Gene ID: | 67836 |
Gene Name: | WD repeat domain containing 83 |
Gene Aliases: |
1500041N16Rik, Morg1 |
Location: |
Chr.8:85075035-85080746 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wdr83 | NM_026399.2 | NP_080675.2 | ||
AK005790.1 | BAB24241.1 | |||
AK020654.1 | BAB32164.1 | |||
AY365434.1 | AAQ64635.1 | |||
BC005587.1 | 3 | 653 | AAH05587.1 | |
BC019369.1 | AAH19369.1 | |||
BC094624.1 | AAH94624.1 |
