Assay Details
Target Gene Details
Entrez Gene ID: | 546049 |
Gene Name: | RIKEN cDNA C330021F23 gene |
Gene Aliases: |
2610027C06Rik, Rps23r1, Rps23rg1 |
Location: |
Chr.8:3567998-3584776 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C330021F23Rik | NM_001024728.2 | NP_001019899.1 | ||
| AK011562.1 | ||||
| AK049309.1 | BAC33674.1 | |||
| AK168456.1 |

Genomic Map