Assay Details
Target Gene Details
Entrez Gene ID: | 11604 |
Gene Name: | agouti related neuropeptide |
Gene Aliases: |
Agrt, Art |
Location: |
Chr.8:105566695-105579845 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agrp | NM_001271806.1 | NP_001258735.1 | ||
| NM_007427.3 | NP_031453.1 | |||
| BC079902.1 | AAH79902.1 | |||
| CO044337.1 |
More Information
Set Membership: |
|

Genomic Map