Assay Details
Target Gene Details
Entrez Gene ID: | 101966 |
Gene Name: | DNA segment, Chr 8, ERATO Doi 738, expressed |
Gene Aliases: |
AA589584 |
Location: |
Chr.8:84246235-84249761 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D8Ertd738e | NM_001007571.2 | NP_001007572.1 | ||
| AK146163.1 | BAE26944.1 | |||
| AK167785.1 | BAE39817.1 | |||
| BC024662.1 | AAH24662.1 | |||
| CA318976.1 | ||||
| CF740495.1 |
Target Gene Details
Entrez Gene ID: | 67873 |
Gene Name: | methylthioribose-1-phosphate isomerase homolog (S. cerevisiae) |
Gene Aliases: |
2410018C20Rik |
Location: |
Chr.8:84249629-84257337 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mri1 |

Genomic Map