Assay Details
Target Gene Details
Entrez Gene ID: | 11499 |
Gene Name: | a disintegrin and metallopeptidase domain 5 |
Gene Aliases: |
tMDC II, tMDCII |
Location: |
Chr.8:24727093-24824369 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam5 | NM_001272057.1 | 16 | 2052 | NP_001258986.1 |
| NM_001272058.1 | 16 | 2052 | NP_001258987.1 | |
| NM_001272059.1 | 16 | 2052 | NP_001258988.1 | |
| NM_007401.3 | 16 | 2052 | NP_031427.2 | |
| AK016574.1 | 16 | 1708 | BAB30315.1 | |
| AK029519.1 | 16 | 1742 | BAC26490.1 | |
| AK161262.1 | 16 | 2052 | BAE36276.1 | |
| AK161415.1 | 13 | 1430 | BAE36383.1 | |
| AK161417.1 | 16 | 1722 | BAE36385.1 | |
| BC094237.1 | 16 | 1749 | AAH94237.1 | |
| U22059.1 | 16 | 1716 | AAA74923.1 |
More Information
Set Membership: |
|

Genomic Map