Assay Details
Target Gene Details
Entrez Gene ID: | 12352 |
Gene Name: | carbonic anhydrase 5a, mitochondrial |
Gene Aliases: |
CAV, Ca5a, Car5 |
Location: |
Chr.8:121916135-121944912 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Car5a | NM_007608.2 | 2 | 299 | NP_031634.2 |
AI098626.1 | 2 | 215 | ||
AI314393.1 | 2 | 299 | ||
AI314492.1 | 2 | 281 | ||
BC030174.1 | 2 | 277 | AAH30174.1 | |
X51971.1 | 2 | 291 | CAA36233.1 |
