Assay Details
Target Gene Details
Entrez Gene ID: | 12352 |
Gene Name: | carbonic anhydrase 5a, mitochondrial |
Gene Aliases: |
CAV, Ca5a, Car5 |
Location: |
Chr.8:121916135-121944912 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Car5a | NM_007608.2 | NP_031634.2 | ||
AI098626.1 | ||||
AI132225.1 | ||||
AI314492.1 | ||||
BC030174.1 | AAH30174.1 | |||
X51971.1 | CAA36233.1 |
