Assay Details
Target Gene Details
Entrez Gene ID: | 74356 |
Gene Name: | RIKEN cDNA 4931428F04 gene |
Gene Aliases: |
AI426165, Kiaa0895l |
Location: |
Chr.8:105280409-105289817 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931428F04Rik | NM_001166394.1 | NP_001159866.1 | ||
| NM_028888.2 | NP_083164.1 | |||
| AK016481.1 | BAB30261.1 | |||
| AK136929.1 | BAE23173.1 | |||
| BC043046.1 | AAH43046.1 | |||
| BC059045.1 | AAH59045.1 |

Genomic Map