Assay Details
Target Gene Details
Entrez Gene ID: | 170786 |
Gene Name: | CD209a antigen |
Gene Aliases: |
CD209, CDSIGN, CIRE, DC-SIGN, DC-SIGN1, Dcsign, SIGN-R1, SIGNR5 |
Location: |
Chr.8:3743395-3748984 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd209a | NM_133238.5 | 3 | 263 | NP_573501.1 |
| AF373408.1 | 3 | 210 | AAL13234.1 | |
| AF374470.1 | 3 | 210 | AAL27539.1 | |
| AK054139.1 | 3 | 227 | BAC35667.1 | |
| AK171639.1 | 3 | 207 | BAE42579.1 | |
| AY049062.1 | 3 | 263 | AAK85825.1 | |
| BC111095.1 | 2 | 311 | AAI11096.1 |
More Information
Set Membership: |
|

Genomic Map