Assay Details
Target Gene Details
Entrez Gene ID: | 52120 |
Gene Name: | heparan-alpha-glucosaminide N-acetyltransferase |
Gene Aliases: |
9430010M12Rik, AW208455, D8Ertd354e, Tmem76 |
Location: |
Chr.8:25944459-25976744 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hgsnat | NM_029884.1 | 10 | 1038 | NP_084160.1 |
| AK035264.1 | 10 | 971 | BAC29006.1 | |
| AK149883.1 | 10 | 1042 | BAE29143.1 | |
| AK152926.1 | 10 | 1003 | BAE31601.1 | |
| AK159649.1 | 10 | 967 | BAE35261.1 | |
| AK160068.1 | 10 | 961 | BAE35603.1 | |
| AK209791.1 | 4 | 304 | ||
| BC024084.1 | 10 | 945 | AAH24084.1 |

Genomic Map