Assay Details
Target Gene Details
Entrez Gene ID: | 52120 |
Gene Name: | heparan-alpha-glucosaminide N-acetyltransferase |
Gene Aliases: |
9430010M12Rik, AW208455, D8Ertd354e, Tmem76 |
Location: |
Chr.8:25944459-25976744 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hgsnat | NM_029884.1 | NP_084160.1 | ||
| AK035264.1 | BAC29006.1 | |||
| AK149883.1 | BAE29143.1 | |||
| AK152926.1 | BAE31601.1 | |||
| AK159649.1 | BAE35261.1 | |||
| AK160068.1 | BAE35603.1 | |||
| AK209731.1 | ||||
| BC024084.1 | AAH24084.1 |

Genomic Map