Assay Details
Target Gene Details
Entrez Gene ID: | 17962 |
Gene Name: | N-acetyltransferase 3 |
Gene Aliases: |
- |
Location: |
Chr.8:67523854-67548627 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nat3 | NM_008674.2 | NP_032700.1 | ||
| AK077364.1 | BAC36769.1 |
More Information
Set Membership: |
|

Genomic Map