Assay Details
Target Gene Details
Entrez Gene ID: | 14232 |
Gene Name: | FK506 binding protein 8 |
Gene Aliases: |
38kDa, FKBP-38, FKBP-8, Fkbp38 |
Location: |
Chr.8:70527743-70535328 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp8 | NM_001111066.1 | NP_001104536.1 | ||
| NM_001199631.1 | NP_001186560.1 | |||
| NM_010223.2 | NP_034353.2 | |||
| AB154247.1 | ||||
| AF030635.1 | AAB86422.1 | |||
| AK088739.1 | BAC40541.1 | |||
| AK146122.1 | BAE26916.1 | |||
| AK152198.1 | BAE31027.1 | |||
| AK153000.1 | ||||
| AK158155.1 | ||||
| AK165281.1 | BAE38118.1 | |||
| AK167663.1 | BAE39713.1 | |||
| AK182349.1 | ||||
| AK197324.1 | ||||
| AK209719.1 | ||||
| AK212094.1 | ||||
| AY225340.1 | AAO39021.1 | |||
| AY278608.1 | AAQ84562.1 | |||
| BC003739.1 | AAH03739.1 | |||
| BC027808.1 | AAH27808.1 |
More Information
Set Membership: |
|

Genomic Map