Assay Details
Target Gene Details
Entrez Gene ID: | 13897 |
Gene Name: | carboxylesterase 1E |
Gene Aliases: |
Eg, Es-22, Es22, egasyn |
Location: |
Chr.8:93201218-93229632 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces1e | NM_133660.3 | NP_598421.1 | ||
| AK144549.1 | BAE25934.1 | |||
| BC019208.1 | AAH19208.1 | |||
| S80191.1 | AAB21335.1 |
More Information
Set Membership: |
|

Genomic Map