Assay Details
Target Gene Details
Entrez Gene ID: | 270096 |
Gene Name: | MON1 homolog b (yeast) |
Gene Aliases: |
5031407H10, 5033413H12Rik |
Location: |
Chr.8:113635586-113645192 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mon1b | NM_001048143.2 | 1 | 61 | NP_001041608.1 |
| AK157390.1 | 1 | 61 | ||
| CJ089261.1 | 1 | 62 |

Genomic Map