Assay Details
Target Gene Details
Entrez Gene ID: | 330817 |
Gene Name: | deoxyhypusine synthase |
Gene Aliases: |
Dhs |
Location: |
Chr.8:85069718-85075162 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dhps | NM_001039514.1 | 2 | 375 | NP_001034603.1 |
AK159104.1 | 2 | 376 | BAE34820.1 |
