Assay Details
Target Gene Details
Entrez Gene ID: | 330817 |
Gene Name: | deoxyhypusine synthase |
Gene Aliases: |
Dhs |
Location: |
Chr.8:85069718-85075162 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dhps | NM_001039514.1 | NP_001034603.1 | ||
AK159104.1 | BAE34820.1 | |||
AK177102.1 | ||||
AK177852.1 | ||||
AK181795.1 | ||||
AK185697.1 | ||||
AK213599.1 | ||||
BC039963.1 | 1 | 226 | AAH39963.1 |
