Assay Details
Target Gene Details
Entrez Gene ID: | 67920 |
Gene Name: | MAK16 homolog (S. cerevisiae) |
Gene Aliases: |
2600016B03Rik, AI314911, Rbm13 |
Location: |
Chr.8:31159468-31168724 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mak16 | NM_026453.3 | NP_080729.1 | ||
| AK015369.1 | BAB29815.1 | |||
| AK075982.1 | BAC36094.1 | |||
| AK075983.1 | BAC36095.1 | |||
| BC013079.1 | AAH13079.1 |

Genomic Map