Assay Details
Target Gene Details
Entrez Gene ID: | 244349 |
Gene Name: | K(lysine) acetyltransferase 6A |
Gene Aliases: |
1500036M03, 9930021N24Rik, MOZ, Myst3, Zfp220 |
Location: |
Chr.8:22859516-22943262 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kat6a | NM_001081149.1 | NP_001074618.1 | ||
| AK028058.1 | BAC25728.1 | |||
| AK036885.1 | BAC29621.1 | |||
| AK082463.1 | ||||
| AK132474.1 | BAE21185.1 | |||
| AK143408.1 | BAE25366.1 | |||
| BC024786.1 |
More Information
Set Membership: |
|

Genomic Map