Assay Details
Target Gene Details
Entrez Gene ID: | 70984 |
Gene Name: | RIKEN cDNA 4931406C07 gene |
Gene Aliases: |
- |
Location: |
Chr.9:15283337-15306448 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4931406C07Rik | NM_001199484.1 | 6 | 708 | NP_001186413.1 |
NM_001199485.1 | 6 | 604 | NP_001186414.1 | |
NM_133732.3 | 5 | 567 | NP_598493.1 | |
AK016432.1 | 5 | 468 | ||
AK088123.1 | 6 | 568 | BAC40160.1 | |
AK133192.1 | 6 | 709 | ||
AK147073.1 | 6 | 578 | BAE27654.1 | |
BC011540.1 | 5 | 470 | AAH11540.1 | |
BC016078.1 | 5 | 516 | AAH16078.1 | |
CT010296.1 | 5 | 381 | CAJ18504.1 |
