Assay Details
Target Gene Details
Entrez Gene ID: | 70984 |
Gene Name: | RIKEN cDNA 4931406C07 gene |
Gene Aliases: |
- |
Location: |
Chr.9:15283337-15306448 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4931406C07Rik | NM_001199484.1 | NP_001186413.1 | ||
NM_001199485.1 | NP_001186414.1 | |||
NM_133732.3 | NP_598493.1 | |||
AK016432.1 | ||||
AK088123.1 | BAC40160.1 | |||
AK133192.1 | ||||
AK147073.1 | BAE27654.1 | |||
BC011540.1 | AAH11540.1 | |||
BC016078.1 | AAH16078.1 | |||
CT010296.1 | CAJ18504.1 |
