Assay Details
Target Gene Details
Entrez Gene ID: | 270086 |
Gene Name: | 2-oxoglutarate and iron-dependent oxygenase domain containing 1 |
Gene Aliases: |
4930415J21Rik, AA387199, AA939912, AW061076, mKIAA1612 |
Location: |
Chr.8:94037141-94067923 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ogfod1 | NM_001093757.1 | 1 | 164 | NP_001087226.1 |
| NM_177767.4 | 1 | 164 | NP_808435.3 | |
| AK029629.1 | 1 | 165 | ||
| AK049824.1 | 1 | 127 | ||
| AK122532.1 | 1 | 71 | BAC65814.1 | |
| AK156765.1 | 1 | 134 | BAE33844.1 | |
| AK164189.1 | 1 | 51 | BAE37671.1 | |
| AK166343.1 | 1 | 87 | BAE38717.1 | |
| BB615031.1 | 1 | 165 | ||
| BC040267.1 | 1 | 124 | AAH40267.1 |
More Information
Set Membership: |
|

Genomic Map