Assay Details
Target Gene Details
Entrez Gene ID: | 270086 |
Gene Name: | 2-oxoglutarate and iron-dependent oxygenase domain containing 1 |
Gene Aliases: |
4930415J21Rik, AA387199, AA939912, AW061076, mKIAA1612 |
Location: |
Chr.8:94037141-94067923 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ogfod1 | NM_001093757.1 | NP_001087226.1 | ||
| NM_177767.4 | NP_808435.3 | |||
| AK029629.1 | ||||
| AK049824.1 | ||||
| AK122532.1 | BAC65814.1 | |||
| AK156765.1 | BAE33844.1 | |||
| AK164189.1 | BAE37671.1 | |||
| AK166343.1 | BAE38717.1 | |||
| BB615031.1 | ||||
| BC040267.1 | AAH40267.1 |
More Information
Set Membership: |
|

Genomic Map