Assay Details
Target Gene Details
Entrez Gene ID: | 20845 |
Gene Name: | steroidogenic acute regulatory protein |
Gene Aliases: |
AV363654, D8Ertd419e |
Location: |
Chr.8:25808513-25815982 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Star | NM_011485.4 | NP_035615.2 | ||
AK014431.1 | ||||
AK019725.2 | BAB31842.1 | |||
AK054470.1 | BAC35791.1 | |||
AK136327.1 | BAE22934.1 | |||
BC060970.1 | AAH60970.1 | |||
BC082283.1 | AAH82283.1 | |||
L36062.1 | AAB94783.1 |
