Assay Details
Target Gene Details
Entrez Gene ID: | 18168 |
Gene Name: | neuropeptide Y receptor Y5 |
Gene Aliases: |
NPY5-R, NPYY5-R, Y5R |
Location: |
Chr.8:66679965-66688581 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npy5r | NM_016708.3 | NP_057917.2 | ||
| AB001346.1 | BAA89538.1 | |||
| BB868845.1 | ||||
| BY008559.1 |
More Information
Set Membership: |
|

Genomic Map